Assay Details
Target Gene Details
Entrez Gene ID: | 29947 |
Gene Name: | DNA methyltransferase 3 like |
Gene Aliases: |
- |
Location: |
Chr.21:44246339-44262216 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNMT3L | NM_013369.3 | NP_037501.2 | ||
| NM_175867.2 | NP_787063.1 | |||
| AF194032.1 | AAF05812.1 | |||
| BC002560.2 | AAH02560.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587742 | Chr.21:44248288 - 44283777 on Build GRCh38 | Loss |
|
| nsv953645 | Chr.21:44156018 - 44264017 on Build GRCh38 | Deletion |
|
| nsv587743 | Chr.21:44255994 - 44263467 on Build GRCh38 | Gain |
|
| esv2723638 | Chr.21:44249174 - 44593709 on Build GRCh38 | Deletion |
|
| nsv587740 | Chr.21:44209682 - 44429981 on Build GRCh38 | Loss |
|
| esv33817 | Chr.21:43749531 - 44647625 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79834782] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map