Assay Details
Target Gene Details
Entrez Gene ID: | 3417 |
Gene Name: | isocitrate dehydrogenase (NADP(+)) 1, cytosolic |
Gene Aliases: |
HEL-216, HEL-S-26, IDCD, IDH, IDP, IDPC, PICD |
Location: |
Chr.2:208236227-208255143 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDH1 | NM_001282386.1 | NP_001269315.1 | ||
| NM_001282387.1 | NP_001269316.1 | |||
| NM_005896.3 | NP_005887.2 | |||
| AF020038.1 | AAD02918.1 | |||
| AF113917.1 | AAD29284.1 | |||
| AK302105.1 | ||||
| AK312242.1 | ||||
| AL136702.1 | CAB66637.1 | |||
| BC012846.1 | AAH12846.1 | |||
| BC093020.1 | AAH93020.1 | |||
| BX537411.1 | CAD97653.1 | |||
| CR533522.1 | CAG38553.1 | |||
| CR536499.1 | CAG38738.1 | |||
| CR541695.1 | CAG46496.1 | |||
| DQ403082.1 | ||||
| EU794623.1 | ||||
| FJ224342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525747 | Chr.2:208222611 - 208252029 on Build GRCh38 | Loss |
|
| nsv584297 | Chr.2:208155457 - 208452035 on Build GRCh38 | Gain |
|
| nsv834517 | Chr.2:208142073 - 208305545 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map