Assay Details
Target Gene Details
Entrez Gene ID: | 5928 |
Gene Name: | RB binding protein 4, chromatin remodeling factor |
Gene Aliases: |
NURF55, RBAP48, lin-53 |
Location: |
Chr.1:32651148-32686211 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP4 | NM_001135255.1 | 12 | 6059 | NP_001128727.1 |
| NM_001135256.1 | 12 | 5962 | NP_001128728.1 | |
| NM_005610.2 | 12 | 6062 | NP_005601.1 | |
| AK056550.1 | 1 | 970 |
Target Gene Details
Entrez Gene ID: | 81493 |
Gene Name: | syncoilin, intermediate filament protein |
Gene Aliases: |
SYNC1, SYNCOILIN |
Location: |
Chr.1:32679906-32702760 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNC | NM_001161708.1 | 3 | 1386 | NP_001155180.1 |
| NM_030786.2 | 3 | 1386 | NP_110413.2 | |
| AK024707.1 | 2 | 461 | BAB14970.1 | |
| AK297958.1 | 3 | 1382 | ||
| AK302275.1 | 3 | 1290 | ||
| BC119700.1 | 2 | 356 | ||
| BC119701.1 | 2 | 356 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461039 | Chr.1:32566811 - 32685350 on Build GRCh38 | Loss |
|
| nsv519188 | Chr.1:32619643 - 32780839 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map