Assay Details
Target Gene Details
Entrez Gene ID: | 1677 |
Gene Name: | DNA fragmentation factor subunit beta |
Gene Aliases: |
CAD, CPAN, DFF-40, DFF2, DFF40 |
Location: |
Chr.1:3857143-3885429 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DFFB | NM_001282669.1 | 7 | 1341 | NP_001269598.1 |
| NM_001320132.1 | 7 | 1438 | NP_001307061.1 | |
| NM_001320136.1 | 7 | 1319 | NP_001307065.1 | |
| NM_004402.3 | 7 | 1269 | NP_004393.1 | |
| NR_104222.1 | 8 | 1406 | ||
| NR_135150.1 | 8 | 1393 | ||
| NR_135151.1 | 8 | 1356 | ||
| NR_135152.1 | 7 | 1291 | ||
| XM_017000498.1 | 6 | 1204 | XP_016855987.1 | |
| XM_017000499.1 | 6 | 1220 | XP_016855988.1 | |
| XM_017000500.1 | 6 | 975 | XP_016855989.1 | |
| AB013918.1 | 7 | 932 | BAA32250.1 | |
| AB028911.1 | 8 | 1186 | BAB40447.1 | |
| AB028912.1 | 7 | 1232 | BAB40448.1 | |
| AB028913.1 | 8 | 1150 | BAB40449.1 | |
| AF039210.1 | 7 | 932 | AAC39709.1 | |
| AF064019.1 | 7 | 1063 | AAC39920.1 | |
| AF409060.1 | 9 | 943 | AAL02005.1 | |
| AF409061.1 | 7 | 958 | AAL02006.1 | |
| AF409062.1 | 7 | 986 | AAL02007.1 | |
| AK290877.1 | 7 | 1060 | ||
| AK303065.1 | 7 | 1148 | ||
| AY438267.1 | 7 | 861 | AAR05821.1 | |
| AY438268.1 | 4 | 484 | AAR05822.1 | |
| BC032827.1 | 8 | 1218 | AAH32827.1 | |
| BC048797.1 | 7 | 1255 | AAH48797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain |
|
| esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain |
|
| nsv509590 | Chr.1:3826457 - 3902492 on Build GRCh38 | Insertion |
|
| esv3585052 | Chr.1:3817921 - 3928224 on Build GRCh38 | Gain |
|
| nsv545130 | Chr.1:3826568 - 4283888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map