Assay Details
Target Gene Details
Entrez Gene ID: | 392 |
Gene Name: | Rho GTPase activating protein 1 |
Gene Aliases: |
CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAP |
Location: |
Chr.11:46677075-46700665 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP1 | NM_004308.3 | 13 | 2085 | NP_004299.1 |
| XM_011520095.1 | 13 | 2522 | XP_011518397.1 | |
| XM_017017720.1 | 11 | 1838 | XP_016873209.1 | |
| AK298562.1 | ||||
| BC018118.1 | 13 | 1990 | AAH18118.1 | |
| U02570.1 | 13 | 1992 | AAA16142.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626191 | Chr.11:46651662 - 46681651 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map