Assay Details
Target Gene Details
Entrez Gene ID: | 84922 |
Gene Name: | FLT3 interacting zinc finger 1 |
Gene Aliases: |
ZNF798 |
Location: |
Chr.19:55591371-55599703 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIZ1 | NM_032836.2 | 3 | 806 | NP_116225.2 |
| XM_005259352.4 | 3 | 1831 | XP_005259409.1 | |
| XM_011527426.2 | 3 | 767 | XP_011525728.1 | |
| AK027674.1 | 3 | 806 | BAB55286.1 | |
| BC132777.1 | 2 | 767 | ||
| BC136512.1 | 2 | 765 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953624 | Chr.19:55302033 - 55625034 on Build GRCh38 | Deletion |
|
| nsv828643 | Chr.19:55547914 - 55643457 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map