Assay Details
Target Gene Details
Entrez Gene ID: | 92797 |
Gene Name: | DNA helicase B |
Gene Aliases: |
DHB, hDHB |
Location: |
Chr.12:66302545-66343643 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HELB | NM_033647.4 | 3 | 739 | NP_387467.2 |
| NR_135080.1 | 3 | 739 | ||
| NR_135081.1 | 3 | 739 | ||
| XM_005269234.2 | 3 | 717 | XP_005269291.1 | |
| XM_017020230.1 | 3 | 717 | XP_016875719.1 | |
| AF319995.1 | 3 | 680 | AAM73554.1 | |
| AK290894.1 | 3 | 704 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629801 | Chr.12:66277937 - 66308122 on Build GRCh38 | Gain |
|
| esv3892212 | Chr.12:66302630 - 66471921 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map