Assay Details
Target Gene Details
Entrez Gene ID: | 11214 |
Gene Name: | A-kinase anchoring protein 13 |
Gene Aliases: |
AKAP-13, AKAP-Lbc, ARHGEF13, BRX, HA-3, Ht31, LBC, PRKA13, PROTO-LB, PROTO-LBC, c-lbc, p47 |
Location: |
Chr.15:85380616-85749358 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKAP13 | NM_006738.5 | NP_006729.4 | ||
| NM_007200.4 | NP_009131.2 | |||
| AB055890.1 | BAB62913.1 | |||
| AK125331.1 | ||||
| BC050312.1 | AAH50312.1 |
Target Gene Details
Entrez Gene ID: | 102465803 |
Gene Name: | microRNA 7706 |
Gene Aliases: |
hsa-mir-7706 |
Location: |
Chr.15:85380596-85380662 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR7706 | NR_106993.1 | |||
| KC832804.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4692n54 | Chr.15:85235254 - 85600273 on Build GRCh38 | Gain |
|
| esv2761699 | Chr.15:85236347 - 85569198 on Build GRCh38 | Gain |
|
| nsv570296 | Chr.15:85237085 - 85383609 on Build GRCh38 | Gain |
|
| esv3637111 | Chr.15:85291627 - 85464874 on Build GRCh38 | Gain |
|
| nsv974618 | Chr.15:85379452 - 85384895 on Build GRCh38 | Duplication |
|
| dgv2648n100 | Chr.15:85268380 - 85605207 on Build GRCh38 | Gain |
|
| esv2751538 | Chr.15:85282336 - 85569198 on Build GRCh38 | Gain |
|
| nsv518064 | Chr.15:85178426 - 85605928 on Build GRCh38 | Gain |
|
| dgv754e212 | Chr.15:85181918 - 85597222 on Build GRCh38 | Gain |
|
| dgv4694n54 | Chr.15:85344022 - 85600273 on Build GRCh38 | Gain |
|
| nsv570298 | Chr.15:85272357 - 85510128 on Build GRCh38 | Gain |
|
| nsv428311 | Chr.15:85107707 - 85486274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map