Assay Details
Target Gene Details
Entrez Gene ID: | 1798 |
Gene Name: | dolichyl-phosphate N-acetylglucosaminephosphotransferase 1 |
Gene Aliases: |
ALG7, CDG-Ij, CDG1J, CMS13, CMSTA2, D11S366, DGPT, DPAGT, DPAGT2, G1PT, GPT, UAGT, UGAT |
Location: |
Chr.11:119096503-119102414 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPAGT1 | NM_001382.3 | 9 | 2006 | NP_001373.2 |
| XM_005271422.2 | 8 | 2100 | XP_005271479.1 | |
| XM_011542648.1 | 7 | 2302 | XP_011540950.1 | |
| XM_017017293.1 | 8 | 2224 | XP_016872782.1 | |
| XM_017017294.1 | 6 | 2077 | XP_016872783.1 | |
| XM_017017295.1 | 7 | 1453 | XP_016872784.1 | |
| AA833740.1 | 1 | 145 | ||
| AK095122.1 | 3 | 2015 | ||
| AK095718.1 | 7 | 2215 | ||
| AK128572.1 | 11 | 3138 | ||
| AK301973.1 | ||||
| BC000325.2 | 9 | 1754 | AAH00325.1 | |
| BM982950.1 | 1 | 154 | ||
| Z82022.1 | 9 | 1665 | CAB04787.1 |
More Information
Set Membership: |
|

Genomic Map