Assay Details
Target Gene Details
Entrez Gene ID: | 8848 |
Gene Name: | TSC22 domain family member 1 |
Gene Aliases: |
Ptg-2, TGFB1I4, TSC22 |
Location: |
Chr.13:44432143-44576565 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1 | NM_001243797.1 | 3 | 473 | NP_001230726.1 |
| NM_001243798.1 | 3 | 478 | NP_001230727.1 | |
| NM_001243799.1 | NP_001230728.1 | |||
| NM_006022.3 | 3 | 815 | NP_006013.1 | |
| NM_183422.3 | 3 | 3797 | NP_904358.2 | |
| AB082525.1 | 3 | 3797 | ||
| AJ222700.1 | 3 | 710 | CAA10951.1 | |
| AK027071.1 | 3 | 2352 | ||
| AK091854.1 | ||||
| AK290751.1 | 3 | 750 | ||
| BC000456.2 | 3 | 731 | AAH00456.1 | |
| BC016867.1 | 3 | 734 | AAH16867.1 | |
| BC069207.1 | 3 | 723 | AAH69207.1 | |
| BC105277.1 | 3 | 726 | AAI05278.1 | |
| BC136475.1 | 3 | 3567 | ||
| BC136482.1 | 3 | 3567 | ||
| BC146664.1 | 3 | 730 | ||
| BC146679.1 | 3 | 730 | ||
| BF307401.1 | 3 | 475 | ||
| BI463370.1 | 3 | 481 | ||
| CN344671.1 | 3 | 453 | ||
| CR627459.1 | 4 | 1990 | CAH10539.1 | |
| U35048.1 | 3 | 701 | AAC50566.1 |
More Information
Set Membership: |
|

Genomic Map