Assay Details
Target Gene Details
Entrez Gene ID: | 84945 |
Gene Name: | abhydrolase domain containing 13 |
Gene Aliases: |
BEM46L1, C13orf6, bA153I24.2 |
Location: |
Chr.13:108218415-108235440 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD13 | NM_032859.2 | 2 | 1619 | NP_116248.2 |
| XM_011521128.2 | 2 | 1577 | XP_011519430.1 | |
| AK027812.1 | 1 | 1076 | BAB55387.1 | |
| AK124864.1 | 1 | 2021 | ||
| BC022566.1 | 2 | 1535 | AAH22566.2 | |
| BC070226.1 | 1 | 1076 | AAH70226.1 | |
| BX647428.1 | 1 | 1199 |
More Information
Set Membership: |
|

Genomic Map