Assay Details
Target Gene Details
Entrez Gene ID: | 25901 |
Gene Name: | coiled-coil domain containing 28A |
Gene Aliases: |
C6orf80, CCRL1AP |
Location: |
Chr.6:138773517-138793319 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC28A | NM_015439.2 | 6 | 1335 | NP_056254.1 |
| AJ420568.1 | 6 | 1086 | ||
| AK300672.1 | 6 | 1086 | ||
| AL050197.1 | 6 | 1085 | CAB43315.1 | |
| AY167571.1 | 6 | 1353 | AAN87344.1 | |
| BC004464.1 | 6 | 1070 | AAH04464.1 | |
| BC013019.2 | 6 | 1067 | ||
| BC080566.1 | 6 | 1069 | AAH80566.1 | |
| BC108717.1 | 6 | 1258 | AAI08718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5500 | Chr.6:138787783 - 138820753 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map