Assay Details
Target Gene Details
Entrez Gene ID: | 9847 |
Gene Name: | C2 calcium dependent domain containing 5 |
Gene Aliases: |
CDP138, KIAA0528 |
Location: |
Chr.12:22448546-22544546 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 25 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759883 | Chr.12:21987900 - 22583973 on Build GRCh38 | Gain+Loss |
|
| nsv557756 | Chr.12:22465671 - 22513597 on Build GRCh38 | Loss |
|
| nsv1038357 | Chr.12:22470636 - 22513887 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map