Assay Details
Target Gene Details
Entrez Gene ID: | 55207 |
Gene Name: | ADP ribosylation factor like GTPase 8B |
Gene Aliases: |
ARL10C, Gie1 |
Location: |
Chr.3:5122245-5180916 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL8B | NM_018184.2 | NP_060654.1 | ||
| AB118751.1 | BAD23992.1 | |||
| AK001564.1 | BAA91759.1 | |||
| AK295465.1 | ||||
| AK298951.1 | ||||
| AK303872.1 | ||||
| BC013131.1 | AAH13131.1 | |||
| BC063125.1 | AAH63125.1 | |||
| CR457264.1 | CAG33545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002376 | Chr.3:5146106 - 5235477 on Build GRCh38 | Gain |
|
| esv3595124 | Chr.3:5138599 - 5176477 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map