Assay Details
Target Gene Details
Entrez Gene ID: | 125488 |
Gene Name: | tetratricopeptide repeat domain 39C |
Gene Aliases: |
C18orf17, HsT2697 |
Location: |
Chr.18:23992773-24135952 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC39C | NM_153211.3 | 1 | 154 | NP_694943.2 |
| XM_017025550.1 | 1 | 84 | XP_016881039.1 | |
| AK091080.1 | 1 | 154 | BAC03578.1 | |
| BC121034.2 | ||||
| BC121035.2 |

Genomic Map