Assay Details
Target Gene Details
Entrez Gene ID: | 25937 |
Gene Name: | WW domain containing transcription regulator 1 |
Gene Aliases: |
TAZ |
Location: |
Chr.3:149517235-149724783 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWTR1 | NM_001168278.1 | 5 | 949 | NP_001161750.1 |
| NM_001168280.1 | 4 | 896 | NP_001161752.1 | |
| NM_015472.4 | 4 | 1032 | NP_056287.1 | |
| XM_017006121.1 | 7 | 1142 | XP_016861610.1 | |
| XM_017006122.1 | 5 | 941 | XP_016861611.1 | |
| AJ299431.1 | 4 | 867 | CAC17722.1 | |
| AK022036.1 | 5 | 949 | BAB13957.1 | |
| AL050107.1 | 2 | 256 | CAB43275.1 | |
| AL833852.1 | 3 | 686 | CAD38711.1 | |
| BC014052.2 | 4 | 926 | AAH14052.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893785 | Chr.3:149507931 - 149583818 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map