Assay Details
Target Gene Details
Entrez Gene ID: | 793 |
Gene Name: | calbindin 1 |
Gene Aliases: |
CALB, D-28K |
Location: |
Chr.8:90058608-90082881 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALB1 | NM_004929.3 | 11 | 1587 | NP_004920.1 |
| AK054881.1 | 11 | 1585 | ||
| AK295585.1 | 10 | 1306 | ||
| AK315959.1 | 10 | 1293 | ||
| BC006478.1 | 11 | 1575 | AAH06478.1 | |
| BX537530.1 | 3 | 4444 | ||
| X06661.1 | 11 | 1426 | CAA29860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759628 | Chr.8:89988558 - 90317441 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map