Assay Details
Target Gene Details
Entrez Gene ID: | 115286 |
Gene Name: | solute carrier family 25 member 26 |
Gene Aliases: |
COXPD28, SAMC |
Location: |
Chr.3:66133610-66380021 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A26 | NM_001164796.1 | NP_001158268.1 | ||
| NM_173471.3 | NP_775742.4 | |||
| NR_028475.1 | ||||
| XM_006712956.1 | XP_006713019.1 | |||
| XM_011533327.2 | XP_011531629.1 | |||
| XM_011533328.1 | XP_011531630.1 | |||
| XM_017005671.1 | XP_016861160.1 | |||
| XM_017005672.1 | XP_016861161.1 | |||
| XM_017005673.1 | XP_016861162.1 | |||
| XM_017005674.1 | XP_016861163.1 | |||
| AJ580932.1 | CAE45652.1 | |||
| AK092495.1 | ||||
| AK096876.1 | ||||
| AK291873.1 | ||||
| AX747571.1 | ||||
| BI822084.1 | ||||
| CR749718.1 | CAI46206.1 | |||
| DA420010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590546 | Chr.3:65487766 - 67232113 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map