Assay Details
Target Gene Details
Entrez Gene ID: | 10632 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit G |
Gene Aliases: |
ATP5JG |
Location: |
Chr.11:118401389-118409847 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5L | NM_006476.4 | 3 | 1164 | NP_006467.4 |
| NR_033759.1 | 4 | 1217 | ||
| AI337729.1 | 1 | 180 | ||
| BC015128.1 | 3 | 952 | AAH15128.1 | |
| BC070165.1 | 3 | 921 | AAH70165.1 |
Target Gene Details
Entrez Gene ID: | 100131626 |
Gene Name: | uncharacterized LOC100131626 |
Gene Aliases: |
- |
Location: |
Chr.11:118381220-118418815 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100131626 | NR_046369.1 | |||
| NR_046370.1 |
More Information
Set Membership: |
|

Genomic Map