Assay Details
Target Gene Details
Entrez Gene ID: | 22797 |
Gene Name: | transcription factor EC |
Gene Aliases: |
TCFEC, TFE-C, TFEC-L, TFECL, bHLHe34, hTFEC-L |
Location: |
Chr.7:115935148-116159891 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TFEC | NM_001018058.2 | 7 | 6052 | NP_001018068.1 |
| NM_001244583.1 | 6 | 5748 | NP_001231512.1 | |
| NM_012252.3 | 8 | 6139 | NP_036384.1 | |
| XM_011515963.1 | 10 | 6326 | XP_011514265.1 | |
| XM_011515964.2 | 10 | 6341 | XP_011514266.1 | |
| XM_011515965.2 | 10 | 6281 | XP_011514267.1 | |
| XM_011515966.2 | 10 | 6390 | XP_011514268.1 | |
| XM_011515969.1 | 9 | 6269 | XP_011514271.1 | |
| XM_011515970.1 | 9 | 6239 | XP_011514272.1 | |
| XM_011515972.2 | 7 | 5855 | XP_011514274.1 | |
| XM_017011874.1 | 9 | 6442 | XP_016867363.1 | |
| XM_017011875.1 | 9 | 6277 | XP_016867364.1 | |
| XM_017011876.1 | 6 | 5714 | XP_016867365.1 | |
| XM_017011877.1 | 8 | 6103 | XP_016867366.1 | |
| XM_017011878.1 | 6 | 5641 | XP_016867367.1 | |
| BX538223.1 | 1 | 4653 | ||
| CR933605.1 | 7 | 6052 | CAI45926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891195 | Chr.7:115561331 - 116511101 on Build GRCh38 | Gain |
|
| nsv831103 | Chr.7:115887640 - 116034159 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map