Assay Details
Target Gene Details
Entrez Gene ID: | 51649 |
Gene Name: | mitochondrial ribosomal protein S23 |
Gene Aliases: |
CGI-138, HSPC329, MRP-S23 |
Location: |
Chr.17:57838926-57850072 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS23 | NM_016070.3 | 5 | 971 | NP_057154.2 |
| AF151896.1 | 5 | 918 | AAD34133.1 | |
| AW299554.1 | ||||
| BI962985.1 |
More Information
Set Membership: |
|

Genomic Map