Assay Details
Target Gene Details
Entrez Gene ID: | 2321 |
Gene Name: | fms related tyrosine kinase 1 |
Gene Aliases: |
FLT, FLT-1, VEGFR-1, VEGFR1 |
Location: |
Chr.13:28300346-28495128 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLT1 | NM_002019.4 | NP_002010.2 | ||
| XM_017020485.1 | XP_016875974.1 | |||
| AB209050.1 | BAD92287.1 | |||
| AF063657.2 | AAC16449.2 | |||
| DQ836394.1 | ||||
| DQ836395.1 | ||||
| EF491868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34052 | Chr.13:28074532 - 28352440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map