Assay Details
Target Gene Details
Entrez Gene ID: | 79809 |
Gene Name: | tetratricopeptide repeat domain 21B |
Gene Aliases: |
ATD4, IFT139, IFT139B, JBTS11, NPHP12, Nbla10696, SRTD4, THM1 |
Location: |
Chr.2:165873362-165953838 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC21B | NM_024753.4 | 4 | 471 | NP_079029.3 |
| XM_006712761.1 | 4 | 414 | XP_006712824.1 | |
| XM_011511872.2 | 4 | 414 | XP_011510174.1 | |
| XM_017004967.1 | 4 | 414 | XP_016860456.1 | |
| XM_017004969.1 | 2 | 114 | XP_016860458.1 | |
| AB073395.1 | 4 | 378 | BAE45724.1 | |
| AK057268.1 | 4 | 409 | BAB71404.1 | |
| AK292942.1 | 4 | 414 | ||
| BC055424.1 | 4 | 471 | AAH55424.1 | |
| DA798655.1 | 2 | 198 |
Target Gene Details
Entrez Gene ID: | 100506134 |
Gene Name: | TTC21B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:165933857-165948321 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC21B-AS1 | NR_038983.1 | |||
| NR_038984.1 | ||||
| AK123487.1 | ||||
| DA373445.1 | ||||
| DA374147.1 | ||||
| DA378622.1 | ||||
| DA379626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834445 | Chr.2:165800387 - 165995152 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map