Assay Details
Target Gene Details
Entrez Gene ID: | 506 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide |
Gene Aliases: |
ATPMB, ATPSB, HEL-S-271 |
Location: |
Chr.12:56638175-56646068 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5B | NM_001686.3 | 9 | 1451 | NP_001677.2 |
| AK291085.1 | 9 | 1367 | ||
| BC016512.1 | 9 | 1356 | AAH16512.1 | |
| DQ403107.1 | 8 | 1154 | ||
| FJ224288.1 | 9 | 1451 | ||
| X05606.1 | 5 | 695 | CAA29095.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map