Assay Details
Target Gene Details
Entrez Gene ID: | 6503 |
Gene Name: | Src-like-adaptor |
Gene Aliases: |
SLA1, SLAP |
Location: |
Chr.8:133036728-133103360 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLA | NM_001045556.2 | 9 | 3350 | NP_001039021.1 |
| NM_001045557.2 | 8 | 3072 | NP_001039022.2 | |
| NM_001282964.1 | 7 | 2940 | NP_001269893.1 | |
| NM_001282965.1 | 7 | 2971 | NP_001269894.1 | |
| NM_006748.3 | 7 | 2716 | NP_006739.2 | |
| XM_017013739.1 | 8 | 3543 | XP_016869228.1 | |
| AW137142.1 | 1 | 126 | ||
| BX648382.1 | 9 | 2933 | ||
| D89077.1 | 7 | 2559 | BAA13758.1 |
Target Gene Details
Entrez Gene ID: | 7038 |
Gene Name: | thyroglobulin |
Gene Aliases: |
AITD3, TGN |
Location: |
Chr.8:132866943-133134902 on Build GRCh38 |
Assay Gene Location: | Within Intron 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TG | NM_003235.4 | NP_003226.4 | ||
| XM_005251038.4 | XP_005251095.1 | |||
| XM_006716622.3 | XP_006716685.1 | |||
| XM_017013793.1 | XP_016869282.1 | |||
| XM_017013794.1 | XP_016869283.1 | |||
| XM_017013795.1 | XP_016869284.1 | |||
| XM_017013796.1 | XP_016869285.1 | |||
| XM_017013797.1 | XP_016869286.1 | |||
| XM_017013798.1 | XP_016869287.1 | |||
| AB209159.1 | BAD92396.1 | |||
| AK299197.1 | ||||
| AK303666.1 | ||||
| AK303667.1 | ||||
| BC127887.1 | ||||
| BC140933.1 | ||||
| U93033.2 | AAC51924.1 | |||
| X05615.1 | CAA29104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482993 | Chr.8:132994656 - 133137835 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map