Assay Details
Target Gene Details
Entrez Gene ID: | 55824 |
Gene Name: | phosphoprotein membrane anchor with glycosphingolipid microdomains 1 |
Gene Aliases: |
CBP, PAG |
Location: |
Chr.8:80967810-81112080 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAG1 | NM_018440.3 | 9 | 9158 | NP_060910.3 |
| XM_017013640.1 | 8 | 9111 | XP_016869129.1 | |
| XM_017013641.1 | 8 | 8710 | XP_016869130.1 | |
| XM_017013642.1 | 8 | 8723 | XP_016869131.1 | |
| XM_017013643.1 | 8 | 8699 | XP_016869132.1 | |
| AK095177.1 | 1 | 1525 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34101 | Chr.8:80912902 - 80985211 on Build GRCh38 | Loss |
|
| nsv396304 | Chr.8:80964174 - 80970124 on Build GRCh38 | Deletion |
|
| nsv831367 | Chr.8:80934819 - 81144000 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map