Assay Details
Target Gene Details
Entrez Gene ID: | 823 |
Gene Name: | calpain 1 |
Gene Aliases: |
CANP, CANP1, CANPL1, SPG76, muCANP, muCL |
Location: |
Chr.11:65181215-65212006 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPN1 | NM_001198868.1 | 11 | 1427 | NP_001185797.1 |
| NM_001198869.1 | 11 | 1396 | NP_001185798.1 | |
| NM_005186.3 | 11 | 1412 | NP_005177.2 | |
| NR_040008.1 | 10 | 1342 | ||
| XM_006718698.1 | 11 | 1327 | XP_006718761.1 | |
| XM_011545292.1 | 11 | 1338 | XP_011543594.1 | |
| AK301538.1 | 10 | 1234 | ||
| AK311001.1 | 11 | 932 | ||
| AK315556.1 | 11 | 1341 | ||
| AY550975.1 | 10 | 1235 | AAT52221.1 | |
| BC008751.2 | 11 | 1363 | AAH08751.1 | |
| BC017200.2 | 11 | 1249 | AAH17200.1 | |
| BC075862.1 | 11 | 1330 | AAH75862.1 | |
| X04366.1 | 11 | 1373 | CAA27881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map