Assay Details
Target Gene Details
Entrez Gene ID: | 687 |
Gene Name: | Kruppel like factor 9 |
Gene Aliases: |
BTEB, BTEB1 |
Location: |
Chr.9:70384597-70414657 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF9 | NM_001206.2 | 2 | 4368 | NP_001197.1 |
| D31716.1 | 2 | 4338 | BAA06524.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain |
|
| nsv831615 | Chr.9:70237789 - 70404776 on Build GRCh38 | Gain |
|
| nsv1049132 | Chr.9:70280466 - 70390981 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map