Assay Details
Target Gene Details
Entrez Gene ID: | 221037 |
Gene Name: | jumonji domain containing 1C |
Gene Aliases: |
TRIP-8, TRIP8 |
Location: |
Chr.10:63167221-63522075 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JMJD1C | NM_001282948.1 | 17 | 6601 | NP_001269877.1 |
| NM_001318153.1 | 17 | 6605 | NP_001305082.1 | |
| NM_001318154.1 | 18 | 6543 | NP_001305083.1 | |
| NM_001322252.1 | 17 | 6598 | NP_001309181.1 | |
| NM_001322254.1 | 16 | 6495 | NP_001309183.1 | |
| NM_001322258.1 | 17 | 6437 | NP_001309187.1 | |
| NM_032776.2 | 18 | 6712 | NP_116165.1 | |
| NR_134512.1 | 14 | 6125 | ||
| XM_011539503.1 | 17 | 6205 | XP_011537805.1 | |
| XM_017015897.1 | 17 | 6200 | XP_016871386.1 | |
| XM_017015898.1 | 18 | 6393 | XP_016871387.1 | |
| XM_017015899.1 | 18 | 6293 | XP_016871388.1 | |
| XM_017015900.1 | 18 | 6697 | XP_016871389.1 | |
| XM_017015901.1 | 17 | 6240 | XP_016871390.1 | |
| XM_017015902.1 | 17 | 6501 | XP_016871391.1 | |
| XM_017015903.1 | 16 | 6049 | XP_016871392.1 | |
| AB037801.1 | 9 | 2552 | BAA92618.1 | |
| AK024991.1 | 7 | 2012 | ||
| AL831917.1 | 11 | 5240 | CAD38578.1 | |
| BC143722.1 | 17 | 6444 | ||
| BX537954.1 | 14 | 6088 | CAD97921.1 | |
| DR762886.1 | ||||
| EF068222.1 | 20 | 6669 | ||
| L40411.1 | 2 | 286 | AAC41741.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677353 | Chr.10:63130283 - 63501631 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map