Assay Details
Target Gene Details
Entrez Gene ID: | 9575 |
Gene Name: | clock circadian regulator |
Gene Aliases: |
KAT13D, bHLHe8 |
Location: |
Chr.4:55427901-55547138 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLOCK | NM_001267843.1 | 24 | 4419 | NP_001254772.1 |
| NM_004898.3 | 23 | 3910 | NP_004889.1 | |
| XM_005265787.2 | 23 | 3956 | XP_005265844.1 | |
| XM_011534409.2 | 23 | 4365 | XP_011532711.1 | |
| XM_011534410.2 | 22 | 3747 | XP_011532712.1 | |
| XM_011534411.2 | 22 | 3706 | XP_011532713.1 | |
| XM_017008854.1 | 24 | 4296 | XP_016864343.1 | |
| XM_017008855.1 | 20 | 3505 | XP_016864344.1 | |
| AB002332.1 | 22 | 3744 | ||
| BX537616.1 | 7 | 1945 |
Target Gene Details
Entrez Gene ID: | 55858 |
Gene Name: | transmembrane protein 165 |
Gene Aliases: |
CDG2K, FT27, GDT1, TMPT27, TPARL |
Location: |
Chr.4:55395913-55453397 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM165 | XM_011534394.2 | XP_011532696.1 |
More Information
Set Membership: |
|

Genomic Map