Assay Details
Target Gene Details
Entrez Gene ID: | 11022 |
Gene Name: | tudor and KH domain containing |
Gene Aliases: |
TDRD2 |
Location: |
Chr.1:151769354-151790576 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDRKH | NM_006862.3 | 14 | 1980 | NP_006853.2 |
| XM_017000123.1 | XP_016855612.1 | |||
| XM_017000126.1 | 14 | 1921 | XP_016855615.1 | |
| XM_017000127.1 | 14 | 1950 | XP_016855616.1 | |
| XM_017000130.1 | 14 | 1814 | XP_016855619.1 | |
| AF119121.2 | 14 | 1968 | AAD30971.1 | |
| AK225160.1 |
More Information
Set Membership: |
|

Genomic Map