Assay Details
Target Gene Details
Entrez Gene ID: | 10973 |
Gene Name: | activating signal cointegrator 1 complex subunit 3 |
Gene Aliases: |
ASC1p200, HELIC1, RNAH |
Location: |
Chr.6:100508194-100881372 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASCC3 | NM_001284271.1 | 1 | 263 | NP_001271200.1 |
| NM_006828.3 | 1 | 263 | NP_006819.2 | |
| NM_022091.4 | 1 | 263 | NP_071374.1 | |
| XM_011535394.2 | 1 | 258 | XP_011533696.1 | |
| XM_011535395.2 | 1 | 252 | XP_011533697.1 | |
| XM_011535396.2 | 1 | 258 | XP_011533698.1 | |
| XM_017010205.1 | 1 | 258 | XP_016865694.1 | |
| XM_017010206.1 | 1 | 255 | XP_016865695.1 | |
| AK299117.1 | 1 | 263 | ||
| AK307903.1 | 1 | 192 | ||
| AK315197.1 | 1 | 196 | ||
| BC026066.1 | 1 | 260 | AAH26066.1 | |
| BC050681.1 | 1 | 164 | AAH50681.1 | |
| BC125211.1 | ||||
| BC125212.1 |
Target Gene Details
Entrez Gene ID: | 102724422 |
Gene Name: | uncharacterized LOC102724422 |
Gene Aliases: |
- |
Location: |
Chr.6:100880867-100896666 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724422 | XM_017011598.1 | XP_016867087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610149 | Chr.6:100750184 - 100963770 on Build GRCh38 | Gain |
|
| esv2763587 | Chr.6:100829998 - 101062615 on Build GRCh38 | Loss |
|
| esv3571321 | Chr.6:100875185 - 100895907 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113405640] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map