Assay Details
Target Gene Details
Entrez Gene ID: | 10739 |
Gene Name: | ret finger protein like 2 |
Gene Aliases: |
RNF79 |
Location: |
Chr.22:32190435-32204844 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFPL2 | NM_001098527.2 | 4 | 1379 | NP_001091997.2 |
| NM_001159545.1 | 3 | 421 | NP_001153017.1 | |
| NM_001159546.1 | 3 | 450 | NP_001153018.1 | |
| NM_006605.3 | 1 | 465 | NP_006596.2 | |
| XM_011529826.1 | 4 | 1583 | XP_011528128.1 | |
| XM_011529827.2 | 5 | 758 | XP_011528129.1 | |
| XM_011529828.1 | 5 | 1493 | XP_011528130.1 | |
| XM_011529830.2 | 5 | 843 | XP_011528132.1 | |
| XM_011529832.1 | 3 | 1469 | XP_011528134.1 | |
| XM_017028535.1 | 4 | 588 | XP_016884024.1 | |
| XM_017028536.1 | 4 | 637 | XP_016884025.1 | |
| AJ010231.1 | 1 | 465 | CAA09045.1 | |
| AK055999.1 | 4 | 1379 | ||
| BC051910.1 | 3 | 421 | AAH51910.1 | |
| BC069737.1 | 1 | 387 | ||
| BC069827.1 | 1 | 386 | ||
| CR456563.1 | 3 | 411 | CAG30449.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575476 | Chr.22:32073030 - 32383792 on Build GRCh38 | Gain |
|
| esv3893474 | Chr.22:32054375 - 32387314 on Build GRCh38 | Gain |
|
| nsv588895 | Chr.22:32178287 - 32470947 on Build GRCh38 | Gain |
|
| nsv834178 | Chr.22:32094462 - 32281534 on Build GRCh38 | Loss |
|
| esv3568364 | Chr.22:32189600 - 32196477 on Build GRCh38 | Loss |
|
| nsv834177 | Chr.22:32020692 - 32198676 on Build GRCh38 | Gain |
|
| nsv966105 | Chr.22:32190542 - 32194436 on Build GRCh38 | Duplication |
|
| esv2422247 | Chr.22:32090461 - 32392271 on Build GRCh38 | Duplication |
|
| nsv3597 | Chr.22:32192437 - 32236737 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map