Assay Details
Target Gene Details
Entrez Gene ID: | 85236 |
Gene Name: | histone cluster 1, H2bk |
Gene Aliases: |
H2B/S, H2BFAiii, H2BFT, H2BK |
Location: |
Chr.6:27138293-27146858 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H2BK | NM_001312653.1 | NP_001299582.1 | ||
| NM_080593.2 | NP_542160.1 | |||
| AJ223352.1 | CAA11276.1 | |||
| AK292062.1 | 1 | 479 | ||
| BC000893.1 | AAH00893.1 | |||
| BC051872.1 | AAH51872.1 | |||
| BC064959.1 | AAH64959.1 | |||
| BC108737.1 | AAI08738.1 | |||
| DA873786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv970091 | Chr.6:27146249 - 27147770 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map