Assay Details
Target Gene Details
Entrez Gene ID: | 26521 |
Gene Name: | translocase of inner mitochondrial membrane 8 homolog B |
Gene Aliases: |
DDP2, TIM8B |
Location: |
Chr.11:112084800-112086798 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIMM8B | NM_012459.2 | NP_036591.2 | ||
| NR_028383.1 | ||||
| AF150087.1 | AAD39994.1 | |||
| AF152350.1 | AAF15100.1 | |||
| AF165967.1 | AAD51801.1 | |||
| AK025397.1 | ||||
| AK312169.1 | ||||
| BC000711.2 | AAH00711.1 | |||
| BC105986.1 | AAI05987.1 | |||
| BC106067.1 | AAI06068.1 | |||
| CF541334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map