Assay Details
Target Gene Details
Entrez Gene ID: | 84708 |
Gene Name: | ligand of numb-protein X 1 |
Gene Aliases: |
LNX, MPDZ, PDZRN2 |
Location: |
Chr.4:53458264-53652524 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX1 | XM_005265785.4 | XP_005265842.1 | ||
| XM_017008776.1 | XP_016864265.1 | |||
| DA874715.1 |
Target Gene Details
Entrez Gene ID: | 100873955 |
Gene Name: | LNX1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.4:53592956-53605382 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX1-AS2 | NR_046827.1 | 2 | 996 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437918 | Chr.4:53585008 - 53617402 on Build GRCh38 | Loss |
|
| nsv829936 | Chr.4:53501616 - 53689956 on Build GRCh38 | Gain |
|
| esv3600530 | Chr.4:53602725 - 53608636 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map