Assay Details
Target Gene Details
Entrez Gene ID: | 101928834 |
Gene Name: | uncharacterized LOC101928834 |
Gene Aliases: |
- |
Location: |
Chr.10:19710335-19728550 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928834 | NR_120646.1 |
Target Gene Details
Entrez Gene ID: | 340895 |
Gene Name: | MAM and LDL receptor class A domain containing 1 |
Gene Aliases: |
C10orf112, DIET1, bA265G8.2 |
Location: |
Chr.10:19046931-19734478 on Build GRCh38 |
Assay Gene Location: | Within Intron 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MALRD1 | NM_001142308.2 | NP_001135780.2 | ||
| XM_011519453.2 | XP_011517755.1 | |||
| XM_011519454.1 | XP_011517756.1 | |||
| XM_011519455.2 | XP_011517757.1 | |||
| XM_017016182.1 | XP_016871671.1 | |||
| XM_017016184.1 | XP_016871673.1 | |||
| XM_017016185.1 | XP_016871674.1 | |||
| XM_017016186.1 | XP_016871675.1 | |||
| AK297683.1 | ||||
| KC843478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039685 | Chr.10:19646166 - 19750692 on Build GRCh38 | Gain |
|
| nsv1049295 | Chr.10:19451367 - 20220107 on Build GRCh38 | Loss |
|
| esv2759737 | Chr.10:19405215 - 19736065 on Build GRCh38 | Loss |
|
| esv3622540 | Chr.10:19536634 - 19746769 on Build GRCh38 | Loss |
|
| nsv831804 | Chr.10:19570471 - 19746664 on Build GRCh38 | Gain |
|
| nsv1046998 | Chr.10:19684167 - 19742539 on Build GRCh38 | Loss |
|
| nsv825266 | Chr.10:19518090 - 19748253 on Build GRCh38 | Loss |
|
| nsv523490 | Chr.10:19686001 - 19752217 on Build GRCh38 | Loss |
|
| esv34306 | Chr.10:19488140 - 19829015 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149975338] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map