Assay Details
Target Gene Details
Entrez Gene ID: | 728215 |
Gene Name: | family with sequence similarity 155 member A |
Gene Aliases: |
- |
Location: |
Chr.13:107168531-107867255 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM155A | NM_001080396.2 | NP_001073865.1 | ||
| XM_011521109.2 | XP_011519411.1 | |||
| AK295445.1 | ||||
| BC146878.1 | ||||
| BC157830.1 | ||||
| BC157855.1 | ||||
| BC157862.1 | ||||
| L10374.1 |
Target Gene Details
Entrez Gene ID: | 100874375 |
Gene Name: | FAM155A intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.13:107787361-107835458 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM155A-IT1 | NR_046848.1 | 2 | 1392 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580880 | Chr.13:107679948 - 107838451 on Build GRCh38 | Loss |
|
| esv2759965 | Chr.13:107639971 - 107824354 on Build GRCh38 | Loss |
|
| nsv1042945 | Chr.13:107707639 - 107788126 on Build GRCh38 | Loss |
|

Genomic Map