Assay Details
Target Gene Details
Entrez Gene ID: | 286430 |
Gene Name: | NLR family pyrin domain containing 2B |
Gene Aliases: |
CLRX.1, NALP2P, NLRP2P, NOD24, POP4 |
Location: |
Chr.X:57677067-57680260 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP2B | NM_001319967.1 | 1 | 1992 | NP_001306896.1 |
| AK026387.1 | 2 | 1491 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv514n21 | Chr.X:57378730 - 57902300 on Build GRCh38 | Gain |
|
| esv2758868 | Chr.X:57605940 - 57863974 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map