Assay Details
Target Gene Details
Entrez Gene ID: | 101927164 |
Gene Name: | uncharacterized LOC101927164 |
Gene Aliases: |
- |
Location: |
Chr.1:224219613-224228043 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927164 | NR_110668.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832692 | Chr.1:224018834 - 224220015 on Build GRCh38 | Gain |
|
| nsv8868 | Chr.1:224195321 - 224564829 on Build GRCh38 | Loss |
|

Genomic Map