Assay Details
Target Gene Details
Entrez Gene ID: | 100313769 |
Gene Name: | microRNA 320b-2 |
Gene Aliases: |
MIR320B-2, MIRN320B2, mir-320b-2 |
Location: |
Chr.1:224257004-224257141 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR320B2 | NR_031574.1 |
Target Gene Details
Entrez Gene ID: | 4931 |
Gene Name: | nuclear VCP-like |
Gene Aliases: |
NVL2 |
Location: |
Chr.1:224227334-224330201 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8868 | Chr.1:224195321 - 224564829 on Build GRCh38 | Loss |
|
| nsv469889 | Chr.1:224252652 - 224446672 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map