Assay Details
Target Gene Details
Entrez Gene ID: | 100852410 |
Gene Name: | ZRANB2 antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.1:71081324-71237723 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZRANB2-AS2 | NR_046217.1 | |||
| BM766420.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577915 | Chr.1:71135255 - 71184361 on Build GRCh38 | Loss |
|
| dgv23e214 | Chr.1:71123716 - 71220341 on Build GRCh38 | Loss |
|
| nsv521769 | Chr.1:71112759 - 71168036 on Build GRCh38 | Loss |
|
| nsv818211 | Chr.1:71138020 - 71168036 on Build GRCh38 | Gain |
|
| esv3586376 | Chr.1:71126870 - 71199890 on Build GRCh38 | Gain |
|

Genomic Map