Assay Details
Target Gene Details
Entrez Gene ID: | 375033 |
Gene Name: | platelet endothelial aggregation receptor 1 |
Gene Aliases: |
JEDI, MEGF12 |
Location: |
Chr.1:156893697-156916434 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PEAR1 | NM_001080471.1 | NP_001073940.1 | ||
| XM_005245141.3 | XP_005245198.1 | |||
| XM_011509510.2 | XP_011507812.1 | |||
| XM_011509511.2 | XP_011507813.1 | |||
| XM_011509512.2 | XP_011507814.1 | |||
| XM_017001236.1 | XP_016856725.1 | |||
| XM_017001237.1 | XP_016856726.1 | |||
| XM_017001238.1 | XP_016856727.1 | |||
| XM_017001239.1 | XP_016856728.1 | |||
| XM_017001242.1 | XP_016856731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss |
|
| esv3890682 | Chr.1:156899922 - 156951428 on Build GRCh38 | Loss |
|
| nsv831659 | Chr.1:156771159 - 156980204 on Build GRCh38 | Loss |
|
| nsv1006558 | Chr.1:156877325 - 156907288 on Build GRCh38 | Gain |
|
| nsv998265 | Chr.1:156834329 - 156903671 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map