Assay Details
Target Gene Details
Entrez Gene ID: | 647219 |
Gene Name: | achaete-scute family bHLH transcription factor 5 |
Gene Aliases: |
bHLHa47 |
Location: |
Chr.1:201113953-201115372 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASCL5 | NM_001270601.1 | 1 | 1078 | NP_001257530.1 |
| AI274185.1 | 1 | 343 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428301 | Chr.1:201059216 - 201191346 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map