Assay Details
Target Gene Details
Entrez Gene ID: | 284498 |
Gene Name: | chromosome 1 open reading frame 167 |
Gene Aliases: |
- |
Location: |
Chr.1:11762188-11789585 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf167 | NM_001010881.1 | 15 | 3246 | NP_001010881.1 |
| XM_011541269.2 | 16 | 3493 | XP_011539571.2 | |
| XM_011541271.2 | 16 | 3288 | XP_011539573.2 | |
| XM_011541272.2 | 16 | 3342 | XP_011539574.1 | |
| XM_011541274.2 | 15 | 3360 | XP_011539576.2 | |
| XM_011541275.2 | 15 | 3157 | XP_011539577.2 | |
| XM_011541276.2 | 16 | 3342 | XP_011539578.1 | |
| XM_011541277.2 | 16 | 3342 | XP_011539579.1 | |
| XM_011541278.2 | 16 | 3342 | XP_011539580.1 | |
| XM_011541280.2 | 11 | 1593 | XP_011539582.2 | |
| XM_011541281.2 | 11 | 1574 | XP_011539583.2 | |
| XM_017001062.1 | 17 | 3480 | XP_016856551.1 | |
| XM_017001063.1 | 16 | 3333 | XP_016856552.1 | |
| XM_017001064.1 | 16 | 3342 | XP_016856553.1 | |
| AL833926.1 | 4 | 562 | CAD38782.1 | |
| AL834308.1 | 8 | 1213 | CAD38978.2 | |
| BC031692.1 | 3 | 392 |

Genomic Map