Assay Details
Target Gene Details
Entrez Gene ID: | 164118 |
Gene Name: | tetratricopeptide repeat domain 24 |
Gene Aliases: |
- |
Location: |
Chr.1:156579727-156587717 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC24 | NM_001105669.2 | NP_001099139.2 | ||
| XM_011509253.2 | XP_011507555.1 | |||
| XM_011509254.2 | XP_011507556.1 | |||
| XM_011509255.2 | XP_011507557.1 | |||
| XM_017000497.1 | XP_016855986.1 | |||
| BQ216668.1 | ||||
| BX414790.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523181 | Chr.1:156580462 - 156706761 on Build GRCh38 | Loss |
|

Genomic Map