Assay Details
Target Gene Details
Entrez Gene ID: | 84838 |
Gene Name: | zinc finger protein 496 |
Gene Aliases: |
NIZP1, ZFP496, ZKSCAN17, ZSCAN49 |
Location: |
Chr.1:247297413-247333376 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833137 | Chr.1:247274015 - 247444126 on Build GRCh38 | Gain |
|
| esv33129 | Chr.1:246911128 - 248165533 on Build GRCh38 | Gain+Loss |
|
| esv3589514 | Chr.1:247297392 - 247354590 on Build GRCh38 | Gain |
|
| nsv509790 | Chr.1:247311000 - 247445876 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map