Assay Details
Target Gene Details
Entrez Gene ID: | 943 |
Gene Name: | TNF receptor superfamily member 8 |
Gene Aliases: |
CD30, D1S166E, Ki-1 |
Location: |
Chr.1:12063318-12144213 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFRSF8 | NM_001243.4 | NP_001234.3 | ||
| NM_001281430.2 | NP_001268359.2 | |||
| XM_011542441.2 | XP_011540743.1 | |||
| BC136400.1 | ||||
| M83554.1 | AAA51947.1 | |||
| S75768.1 | AAD14188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525898 | Chr.1:12081404 - 12108466 on Build GRCh38 | Loss |
|
| nsv469830 | Chr.1:12039669 - 12199858 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150060465] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map