Assay Details
Target Gene Details
Entrez Gene ID: | 64769 |
Gene Name: | MYST/Esa1 associated factor 6 |
Gene Aliases: |
C1orf149, CENP-28, EAF6, NY-SAR-91 |
Location: |
Chr.1:37489960-37514819 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEAF6 | NM_001270875.1 | 7 | 2893 | NP_001257804.1 |
| NM_001270876.1 | 6 | 2566 | NP_001257805.1 | |
| NM_022756.5 | 8 | 2923 | NP_073593.2 | |
| NR_073090.1 | 9 | 2981 | ||
| NR_073091.1 | 8 | 2951 | ||
| NR_073092.1 | 9 | 3012 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519551 | Chr.1:37491775 - 37509678 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map