Assay Details
Target Gene Details
Entrez Gene ID: | 374973 |
Gene Name: | testis expressed 38 |
Gene Aliases: |
ATPAF1-AS1, C1orf223, THEG4 |
Location: |
Chr.1:46669344-46673603 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEX38 | NM_001145474.3 | 2 | 198 | NP_001138946.1 |
| NM_001300863.1 | NP_001287792.1 | |||
| NM_001300864.1 | NP_001287793.1 | |||
| XM_011541421.2 | 2 | 194 | XP_011539723.1 | |
| BC057818.1 | 2 | 177 | AAH57818.1 | |
| BC126318.1 | 2 | 153 | ||
| BC144026.1 | ||||
| BC144027.1 | ||||
| FJ211596.1 | 2 | 96 |
More Information
Set Membership: |
|

Genomic Map